| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800005269.mm.1 |
NM_172631.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005269.mm.1 |
NM_172631.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005269.mm.1 |
TC1800000714.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005270.mm.1 |
NM_172631.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005270.mm.1 |
NM_172631.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005270.mm.1 |
TC1800000714.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005271.mm.1 |
NM_172631.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005271.mm.1 |
NM_172631.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005271.mm.1 |
TC1800000714.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005273.mm.1 |
NM_172631.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005273.mm.1 |
NM_172631.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005273.mm.1 |
TC1800000714.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005272.mm.1 |
NM_172631.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005272.mm.1 |
NM_172631.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005272.mm.1 |
TC1800000714.mm.1 |
MTA TC |