| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800005426.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005426.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005426.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005426.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005426.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005424.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005424.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005424.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005424.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005424.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005427.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005427.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005427.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005427.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005427.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005420.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005420.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005420.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005420.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005420.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005417.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005417.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005417.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005417.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005417.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005421.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005421.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005421.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005421.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005421.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005422.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005422.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005422.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005422.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005422.mm.1 |
TC1800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800005416.mm.1 |
NM_001083967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005416.mm.1 |
NM_001083967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005416.mm.1 |
NM_013685.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005416.mm.1 |
NM_013685.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005416.mm.1 |
TC1800000741.mm.1 |
MTA TC |