Assay Details
| Gene Symbol: Poli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136090.2 | NP_001129562.1 | 1 - 2 | 575 | 60 | |
| NM_001289515.1 | NP_001276444.1 | 1 - 2 | 575 | 60 | ||
| NM_001289516.1 | NP_001276445.1 | 1 - 2 | 575 | 60 | ||
| GenBank mRNA | AK154537.1 | - | 1 - 2 | 524 | 60 | |
| BC057575.1 | - | 1 - 2 | 535 | 60 | ||
| BC082278.1 | - | 1 - 2 | 392 | 6328261 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154537.1, BC082278.1, NM_001136090.2, NM_001289515.1, NM_001289516.1 transcripts.
Set Membership: |
|

Poli
Genomic Map