Assay Details
| Gene Symbol: Poli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136090.2 | NP_001129562.1 | 9 - 10 | 1848 | 61 | |
| NM_001289515.1 | NP_001276444.1 | 9 - 10 | 2493 | 61 | ||
| NM_001289516.1 | NP_001276445.1 | 9 - 10 | 1779 | 61 | ||
| NM_011972.2 | NP_036102.2 | 9 - 10 | 1411 | 61 | ||
| GenBank mRNA | AF151691.1 | - | 9 - 10 | 1411 | 61 | |
| AK154537.1 | - | 9 - 10 | 2442 | 61 | ||
| AY515316.1 | - | 9 - 10 | 1317 | 61 | ||
| BC057575.1 | - | 9 - 10 | 1739 | 61 | ||
| BC082278.1 | - | 9 - 10 | 1665 | 6328261 | 61 | |
| HQ698885.1 | - | 9 - 10 | 1544 | 61 | ||
| HQ698886.1 | - | 9 - 10 | 1544 | 61 | ||
| HQ698887.1 | - | 9 - 10 | 1544 | 61 | ||
| HQ698889.1 | - | 9 - 10 | 1544 | 61 | ||
| HQ698890.1 | - | 9 - 10 | 1544 | 61 | ||
| HQ698892.1 | - | 9 - 10 | 1544 | 61 | ||
| HQ698893.1 | - | 9 - 10 | 1544 | 61 | ||
More Information
Set Membership: |
|

Poli
Genomic Map