Assay Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008292.4 | NP_032318.2 | 7 - 8 | 576 | 98 | |
| GenBank mRNA | AK004866.1 | - | 7 - 8 | 576 | 98 | |
| AK088381.1 | - | 7 - 8 | 551 | 98 | ||
| AK147182.1 | - | 7 - 8 | 548 | 98 | ||
| AK152532.1 | - | 7 - 8 | 471 | 98 | ||
| AK161655.1 | - | 7 - 8 | 471 | 98 | ||
| AK166801.1 | - | 7 - 8 | 489 | 98 | ||
| AK167060.1 | - | 7 - 8 | 526 | 98 | ||
| AK169077.1 | - | 7 - 8 | 478 | 98 | ||
| AK199910.1 | - | 1 - 2 | 32 | 98 | ||
| AK214272.1 | - | 1 - 2 | 33 | 98 | ||
| BC022175.1 | - | 7 - 8 | 449 | 4209394 | 98 | |
| X89998.1 | - | 7 - 8 | 437 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b4
Genomic Map