Assay Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008292.4 | NP_032318.2 | 14 - 15 | 1408 | 82 | |
| GenBank mRNA | AK004866.1 | - | 14 - 15 | 1408 | 82 | |
| AK088381.1 | - | 14 - 15 | 1383 | 82 | ||
| AK147182.1 | - | 14 - 15 | 1380 | 82 | ||
| AK152532.1 | - | 14 - 15 | 1303 | 82 | ||
| AK161655.1 | - | 14 - 15 | 1303 | 82 | ||
| AK166801.1 | - | 14 - 15 | 1321 | 82 | ||
| AK167060.1 | - | 14 - 15 | 1358 | 82 | ||
| AK169077.1 | - | 14 - 15 | 1310 | 82 | ||
| AK187517.1 | - | 2 - 3 | 299 | 82 | ||
| BC022175.1 | - | 14 - 15 | 1281 | 4209394 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b4
Genomic Map