Assay Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008292.4 | NP_032318.2 | 15 - 16 | 1481 | 96 | |
| GenBank mRNA | AK004866.1 | - | 15 - 16 | 1481 | 96 | |
| AK088381.1 | - | 15 - 16 | 1456 | 96 | ||
| AK147182.1 | - | 15 - 16 | 1453 | 96 | ||
| AK152532.1 | - | 15 - 16 | 1376 | 95 | ||
| AK161655.1 | - | 15 - 16 | 1376 | 96 | ||
| AK166801.1 | - | 15 - 16 | 1394 | 96 | ||
| AK167060.1 | - | 15 - 16 | 1431 | 96 | ||
| AK169077.1 | - | 15 - 16 | 1383 | 96 | ||
| BC022175.1 | - | 15 - 16 | 1354 | 4209394 | 96 | |
| X89998.1 | - | 15 - 16 | 1342 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b4
Genomic Map