Assay Details
| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY350709.1 | - | 1 - 2 | 101 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY350709.1 transcripts.
Set Membership: |
|

Zfp474
Genomic Map