Assay Details
| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY350709.1 | - | 3 - 4 | 350 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY350709.1 transcripts.
Set Membership: |
|

Zfp474
Genomic Map