Assay Details
| Gene Symbol: Eif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010120.5 | NP_034250.3 | 1 - 2 | 94 | 92 | |
| GenBank mRNA | AK009026.1 | - | 1 - 2 | 65 | 92 | |
| AK013268.1 | - | 1 - 2 | 93 | 92 | ||
| AK019451.1 | - | 1 - 2 | 95 | 92 | ||
| AK150651.1 | - | 1 - 2 | 91 | 92 | ||
| AK150673.1 | - | 1 - 2 | 67 | 92 | ||
| AK150677.1 | - | 1 - 2 | 67 | 92 | ||
| AK167333.1 | - | 1 - 2 | 54 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009026.1, AK013268.1, AK019451.1, AK150651.1, AK150673.1, AK150677.1, AK167333.1, NM_010120.5 transcripts.
Set Membership: |
|

Eif1a
Genomic Map