Assay Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291455.1 | NP_001278384.1 | 1 - 2 | 456 | 78 | |
| GenBank mRNA | AF501324.1 | - | 1 - 2 | 407 | 78 | |
More Information
Set Membership: |
|

Dpysl3
Genomic Map