Assay Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136086.2 | NP_001129558.1 | 2 - 3 | 298 | 117 | |
| NM_001291455.1 | NP_001278384.1 | 2 - 3 | 548 | 117 | ||
| NM_009468.5 | NP_033494.1 | 2 - 3 | 479 | 117 | ||
| GenBank mRNA | AF501324.1 | - | 2 - 3 | 499 | 117 | |
| AK077000.1 | - | 2 - 3 | 289 | 117 | ||
| AK133040.1 | - | 2 - 3 | 298 | 117 | ||
| AK149024.1 | - | 2 - 3 | 317 | 117 | ||
| AK161509.1 | - | 2 - 3 | 220 | 117 | ||
| BC023003.1 | - | 2 - 3 | 163 | 5367381 | 117 | |
| X87817.1 | - | 2 - 3 | 439 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl3
Genomic Map