Assay Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136086.2 | NP_001129558.1 | 9 - 10 | 1135 | 66 | |
| NM_001291455.1 | NP_001278384.1 | 9 - 10 | 1385 | 66 | ||
| NM_009468.5 | NP_033494.1 | 9 - 10 | 1316 | 66 | ||
| GenBank mRNA | AK077000.1 | - | 9 - 10 | 1126 | 66 | |
| AK133040.1 | - | 9 - 10 | 1135 | 66 | ||
| AK149024.1 | - | 9 - 10 | 1153 | 66 | ||
| AK161509.1 | - | 9 - 10 | 1057 | 66 | ||
| AK171653.1 | - | 2 - 3 | 216 | 66 | ||
| BC023003.1 | - | 9 - 10 | 1000 | 5367381 | 66 | |
| X87817.1 | - | 9 - 10 | 1276 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl3
Genomic Map