Assay Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136086.2 | NP_001129558.1 | 1 - 2 | 211 | 117 | |
| GenBank mRNA | AK077000.1 | - | 1 - 2 | 202 | 117 | |
| AK133040.1 | - | 1 - 2 | 211 | 117 | ||
| AK161509.1 | - | 1 - 2 | 133 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133040.1, AK161509.1, NM_001136086.2 transcripts.
Set Membership: |
|

Dpysl3
Genomic Map