Assay Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152809.2 | NP_690022.2 | 3 - 4 | 957 | 102 | |
| GenBank mRNA | CN454971.1 | - | 2 - 3 | 440 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1g3
Genomic Map