Assay Details
| Gene Symbol: Arhgef37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177828.4 | NP_808496.2 | 3 - 4 | 465 | 101 | |
| GenBank mRNA | AB199795.1 | - | 2 - 3 | 315 | 101 | |
| CJ131942.1 | - | 3 - 4 | 366 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef37
Genomic Map