Assay Details
| Gene Symbol: Orm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008768.1 | NP_032794.1 | 2 - 3 | 291 | 73 | |
| GenBank mRNA | BC012725.1 | - | 2 - 3 | 298 | 4162961 | 73 |
| CT010399.1 | - | 2 - 3 | 259 | 73 | ||
| M27008.1 | - | 2 - 3 | 291 | 73 | ||
More Information
Set Membership: |
|

Orm1
Genomic Map