Assay Details
Gene Symbol: Pcdhgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033575.3 | NP_291053.1 | 1 - 2 | 2510 | 73 | |
GenBank mRNA | AF464151.1 | - | 1 - 2 | 151 | 73 | |
AF464152.1 | - | 1 - 2 | 151 | 73 | ||
AY013805.1 | - | 1 - 2 | 2411 | 73 | ||
BC054557.1 | - | 1 - 2 | 2510 | 5706538 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
