Assay Details
| Gene Symbol: Pcdhb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053142.3 | NP_444372.2 | 1 | 3190 | 154 | |
| GenBank mRNA | AF326310.1 | - | 1 | 3052 | 154 | |
| AK077255.1 | - | 1 | 3161 | 154 | ||
| AK122533.1 | - | 3416 | 154 | |||
| BC017149.1 | - | 1 | 3200 | 4506788 | 154 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175937 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107051 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180741 | BC017149.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180741 | AF326310.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180741 | AK077255.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180741 | AK122533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180741 | NM_053142.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628523 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08968294 | - | - |
| Whole Mouse Genome G4122A | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | BC017149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | AF326310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | AK077255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | AK122533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P337771 | NM_053142.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180741 | BC017149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180741 | AF326310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180741 | AK077255.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180741 | AK122533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180741 | NM_053142.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14009124 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14009145 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14009083 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14009147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13821107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14009155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14009160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14009083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628533 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455115 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448933_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455112 | - | - |
| Mouse Genome 430 2.0 Array | 1448933_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448933_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93888_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93888_at | - | - |
| Mouse Expression Set 430 | 1448933_PM_at | - | - |
| Mouse Expression Set 430 | 1448933_at | - | - |
| Murine Genome U74v2 Set | 168300_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168390_f_at | - | - |
| Murine Genome U74v2 Set | 168390_PM_f_at | - | - |
| Murine Genome U74v2 Set | 168300_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448933_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448933_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448933_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448933_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 752340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214296 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214296 | - | - |
| mouse-6 v1.1 (Illumina) | scl093888.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl093888.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214296 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214296 | - | - |

Pcdhb17
Genomic Map