Assay Details
Gene Symbol: Pcdhb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053139.3 | NP_444369.3 | 1 | 2532 | 116 | |
GenBank mRNA | AK032156.1 | - | 1 | 2535 | 116 | |
AK083065.1 | - | 1 | 2481 | 116 | ||
AY013775.1 | - | 1 | 2346 | 116 | ||
BC029681.1 | - | 1 | 1914 | 116 | ||
BC059821.1 | - | 1 | 2509 | 6838626 | 116 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | BC059821.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | BC029681.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | AK032156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | AK083065.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | BC059821.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | BC029681.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | AK032156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | AK083065.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173090 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662985 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149668 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | BC059821.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | BC029681.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | AK032156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | AK083065.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | BC059821.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | BC029681.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | AK032156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | AK083065.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | BC059821.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | BC029681.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | AK032156.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | AK083065.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | BC059821.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | BC029681.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | AK032156.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | AK083065.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628328 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628339 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628330 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628349 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628345 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628351 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149668 | - | - |
Whole Mouse Genome G4122A | A_51_P240421 | BC059821.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | BC029681.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | AK032156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | AK083065.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | BC059821.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | BC029681.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | AK032156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | AK083065.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P240421 | NM_053139.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134289 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149668 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019134 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019150 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019122 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019119 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019168 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019152 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14019138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628371 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628346 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628349 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14019165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628345 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13917281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628356 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13986716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13986715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628379 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628371 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628382 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628354 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14019150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628378 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628420 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628372 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628375 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14019167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628335 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10455102 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455103 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455099 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455101 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10455100 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421437_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421438_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10455098 | - | - |
Mouse Genome 430 2.0 Array | 1421437_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1421438_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1421437_x_at | - | - |
Mouse Genome 430 2.0 Array | 1421438_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93885_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93885_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800002849.mm.1 | NM_053139.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002849.mm.1 | NM_053139.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800002849.mm.1 | TC1800000370.mm.1 | MTA TC |
Mouse Expression Set 430 | 1421438_at | - | - |
Mouse Expression Set 430 | 1421437_PM_x_at | - | - |
Mouse Expression Set 430 | 1421437_x_at | - | - |
Mouse Expression Set 430 | 1421438_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421437_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1421437_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1421438_at | - | - |
Mouse Genome 430A 2.0 Array | 1421438_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421437_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421438_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421437_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421438_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 641040 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2370411 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002370411 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2607000 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2607000 | - | - |
mouse-6 v1.1 (Illumina) | scl093885.1_11-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2370411 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl093885.1_11-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370411 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2607000 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2607000 | - | - |
