Assay Details
| Gene Symbol: Pcdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028374.1 | - | 1 - 2 | 327 | 80 | |
| BC127076.1 | - | 1 - 2 | 225 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC127076.1 transcripts.
Set Membership: |
|

Pcdh12
Genomic Map