Assay Details
Gene Symbol: Cep120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178686.3 | NP_848801.2 | 13 - 14 | 1939 | 87 | |
GenBank mRNA | AK038916.1 | - | 13 - 14 | 2001 | 87 | |
AK044808.1 | - | 12 - 13 | 3004 | 87 | ||
AK079319.1 | - | 13 - 14 | 1956 | 87 | ||
AK089236.1 | - | 13 - 14 | 1946 | 87 | ||
AK089242.1 | - | 13 - 14 | 1946 | 87 | ||
BC046493.1 | - | 5 - 6 | 805 | 87 | ||
BC053439.1 | - | 13 - 14 | 1939 | 6328829 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
