Assay Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK160482.1 | - | 1 - 2 | 85 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160482.1 transcripts.
Set Membership: |
|

Echdc2
Genomic Map