Assay Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282017.1 | NP_001268946.1 | 4 - 5 | 450 | 103 | |
| NM_008951.2 | NP_032977.1 | 4 - 5 | 450 | 103 | ||
| GenBank mRNA | AB029142.1 | - | 4 - 5 | 381 | 103 | |
| AB029143.1 | - | 4 - 5 | 381 | 103 | ||
| AB029144.1 | - | 4 - 5 | 379 | 103 | ||
| AB029145.1 | - | 4 - 5 | 381 | 103 | ||
| AB029146.1 | - | 4 - 5 | 369 | 103 | ||
| AF013099.1 | - | 4 - 5 | 381 | 103 | ||
| AF175574.1 | - | 4 - 5 | 381 | 103 | ||
| AK010969.1 | - | 4 - 5 | 379 | 103 | ||
| AK145524.1 | - | 4 - 5 | 389 | 103 | ||
| BC009005.1 | - | 4 - 5 | 399 | 3581937 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmd4
Genomic Map