Assay Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282017.1 | NP_001268946.1 | 9 - 10 | 1046 | 65 | |
| NM_008951.2 | NP_032977.1 | 9 - 10 | 1037 | 65 | ||
| GenBank mRNA | AB029142.1 | - | 9 - 10 | 968 | 65 | |
| AB029143.1 | - | 9 - 10 | 977 | 65 | ||
| AF013099.1 | - | 9 - 10 | 968 | 65 | ||
| AF175574.1 | - | 9 - 10 | 977 | 65 | ||
| AK010969.1 | - | 9 - 10 | 966 | 65 | ||
| AK145524.1 | - | 9 - 10 | 985 | 65 | ||
| BC009005.1 | - | 9 - 10 | 995 | 3581937 | 65 | |
| BC016433.1 | - | 6 - 7 | 1836 | 65 | ||
| BY473933.1 | - | 1 - 2 | 45 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341987 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341989 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990894 | AK010969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990894 | AK145524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990894 | BC009005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990894 | BC016433.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990894 | AF013099.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28834 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990894 | AK010969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990894 | AK145524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990894 | BC009005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990894 | BC016433.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990894 | AF013099.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P12869 | AB029145.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P12869 | AB029144.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P207962 | AK010969.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P207962 | AK145524.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P207962 | BC009005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P207962 | BC016433.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P207962 | AB029145.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341998 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04994023 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04994035 | - | - |
| Whole Mouse Genome G4122A | A_52_P12869 | AB029145.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P12869 | AB029144.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207962 | AK010969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207962 | AK145524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207962 | BC009005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207962 | BC016433.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207962 | AB029145.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02737740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02737741 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02737737 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02737746 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341987 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02567844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02406769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02725379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02511508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02284886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03169895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02468333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02567845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02511512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500060 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451725_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418874_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425859_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500054 | - | - |
| Mouse Genome 430 2.0 Array | 1418874_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425859_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451725_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418874_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451725_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425859_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19185_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19185_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018964.mm.1 | NM_001282017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018964.mm.1 | NM_008951.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018964.mm.1 | TC0300002462.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018966.mm.1 | NM_001282017.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018966.mm.1 | NM_008951.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018966.mm.1 | TC0300002462.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451725_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451725_a_at | - | - |
| Mouse Expression Set 430 | 1425859_a_at | - | - |
| Mouse Expression Set 430 | 1418874_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425859_PM_a_at | - | - |
| Mouse Expression Set 430 | 1418874_a_at | - | - |
| Murine Genome U74v2 Set | 94302_at | - | - |
| Murine Genome U74v2 Set | 94302_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418874_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425859_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451725_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451725_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418874_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425859_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418874_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451725_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451725_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418874_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425859_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425859_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 430068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106760528 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 430068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000430068 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228630 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2449651 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228630 | - | - |
| mouse-6 v1.1 (Illumina) | scl21852.5_6-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001959.1_79-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 430068 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106760528 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21852.5_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106760528 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430068 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228630 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228630 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21852.5_6-S | - | - |

Psmd4
Genomic Map