| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_001042418.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_001042418.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_001042419.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_001042419.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_001042420.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_001042420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_027687.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_027687.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_181731.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
NM_181731.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000824.mm.1 |
TC1800000107.mm.1 |
MTA TC |