Assay Details
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042418.1 | NP_001035883.1 | 4 - 5 | 1440 | 74 | |
| GenBank mRNA | AF359385.1 | - | 4 - 5 | 1440 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cabyr
Genomic Map