Assay Details
| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008945.3 | NP_032971.2 | 3 - 4 | 548 | 66 | |
| GenBank mRNA | AK077448.1 | - | 3 - 4 | 525 | 66 | |
| AK137044.1 | - | 3 - 4 | 524 | 66 | ||
| AK156624.1 | - | 3 - 4 | 524 | 66 | ||
| AK161348.1 | - | 3 - 4 | 524 | 66 | ||
| AK168265.1 | - | 3 - 4 | 524 | 66 | ||
| AK168340.1 | - | 3 - 4 | 524 | 66 | ||
| AK194462.1 | - | 2 - 3 | 253 | 66 | ||
| AK204169.1 | - | 1 - 2 | 66 | 66 | ||
| BC008241.1 | - | 3 - 4 | 503 | 2654421 | 66 | |
| CB058553.1 | - | 3 - 4 | 548 | 66 | ||
| U65636.1 | - | 3 - 4 | 503 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb4
Genomic Map