Assay Details
| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008945.3 | NP_032971.2 | 4 - 5 | 626 | 62 | |
| GenBank mRNA | AK077448.1 | - | 4 - 5 | 603 | 62 | |
| AK137044.1 | - | 4 - 5 | 602 | 62 | ||
| AK156624.1 | - | 4 - 5 | 602 | 62 | ||
| AK161348.1 | - | 4 - 5 | 602 | 62 | ||
| AK168265.1 | - | 4 - 5 | 602 | 62 | ||
| AK168340.1 | - | 4 - 5 | 602 | 62 | ||
| AK194462.1 | - | 3 - 4 | 331 | 62 | ||
| AK204169.1 | - | 2 - 3 | 144 | 62 | ||
| BC008241.1 | - | 4 - 5 | 581 | 2654421 | 62 | |
| CB058553.1 | - | 4 - 5 | 626 | 62 | ||
| U65636.1 | - | 4 - 5 | 581 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of inflammatory response to antigenic stimulus
proteolysis
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
lipopolysaccharide binding
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
negative regulation of inflammatory response to antigenic stimulus
proteolysis
proteasomal ubiquitin-independent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
lipopolysaccharide binding
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341206 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341204 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341207 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341203 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | U65636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | U65636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803435 | AK003830.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803435 | BC055880.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803435 | NM_026753.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126574 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123783 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | U65636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | U65636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | U65636.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | U65636.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22341196 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123783 | - | - |
| Whole Mouse Genome G4122A | A_51_P155522 | U65636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | U65636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK168265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK168340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK156624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155522 | AK161348.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134340 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114114 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123783 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02338346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02338348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02937873 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02937880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341207 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341205 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341203 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341206 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22341204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22341191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02937873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03057369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02937876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02796904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02937875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02338348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02338346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02937877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02550011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02937880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02796901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02475934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22341216 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500036 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500040 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500039 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500037 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500038 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500035 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438984_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451205_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500034 | - | - |
| Mouse Genome 430 2.0 Array | 1438984_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438984_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451205_at | - | - |
| Mouse Genome 430 2.0 Array | 1451205_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19172_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19172_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018884.mm.1 | NM_008945.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018884.mm.1 | NM_008945.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018884.mm.1 | TC0300002454.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438984_x_at | - | - |
| Mouse Expression Set 430 | 1438984_PM_x_at | - | - |
| Mouse Expression Set 430 | 1451205_at | - | - |
| Mouse Expression Set 430 | 1451205_PM_at | - | - |
| Murine Genome U74v2 Set | 98556_at | - | - |
| Murine Genome U74v2 Set | 98557_PM_f_at | - | - |
| Murine Genome U74v2 Set | 98557_f_at | - | - |
| Murine Genome U74v2 Set | 98556_PM_at | - | - |
| Murine Genome U74v2 Set | 98558_r_at | - | - |
| Murine Genome U74v2 Set | 98558_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438984_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451205_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451205_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438984_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438984_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451205_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451205_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438984_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 636072 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 520402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 520402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671124 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2671124 | - | - |
| mouse-6 v1.1 (Illumina) | scl21857.5.1_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21857.5.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2671124 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671124 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21857.5.1_13-S | - | - |

Psmb4
Genomic Map