Assay Details
| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008945.3 | NP_032971.2 | 5 - 6 | 746 | 63 | |
| GenBank mRNA | AK137044.1 | - | 5 - 6 | 722 | 63 | |
| AK168265.1 | - | 5 - 6 | 722 | 63 | ||
| AK168340.1 | - | 5 - 6 | 722 | 63 | ||
| BC008241.1 | - | 5 - 6 | 701 | 2654421 | 63 | |
| U65636.1 | - | 5 - 6 | 701 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb4
Genomic Map