Assay Details
| Gene Symbol: Scnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163573.1 | NP_001157045.1 | 2 - 3 | 239 | 75 | |
| NM_027013.2 | NP_081289.2 | 2 - 3 | 239 | 75 | ||
| GenBank mRNA | AK013948.1 | - | 2 - 3 | 240 | 75 | |
| BC028867.1 | - | 2 - 3 | 215 | 4220000 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scnm1
Genomic Map