Assay Details
| Gene Symbol: Scnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163573.1 | NP_001157045.1 | 4 - 5 | 419 | 67 | |
| NM_027013.2 | NP_081289.2 | 4 - 5 | 419 | 67 | ||
| GenBank mRNA | AK013948.1 | - | 4 - 5 | 420 | 67 | |
| BC028867.1 | - | 4 - 5 | 395 | 4220000 | 67 | |
| BQ032998.1 | - | 2 - 3 | 67 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113155 | NM_027013.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113155 | AK013948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113155 | BC028867.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113155 | BU756175.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003581 | NM_001163573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003581 | NM_027013.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003581 | BC028867.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003581 | BQ032998.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003581 | BU756175.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167192 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176523 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003581 | NM_001163573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003581 | NM_027013.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003581 | BC028867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003581 | BQ032998.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003581 | BU756175.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113155 | NM_027013.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113155 | AK013948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113155 | BC028867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113155 | BU756175.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284597 | NM_001163573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284597 | NM_027013.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284597 | AK013948.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284597 | BC028867.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284597 | BQ032998.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342387 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176523 | - | - |
| Whole Mouse Genome G4122A | A_51_P284597 | NM_001163573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284597 | NM_027013.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284597 | AK013948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284597 | BC028867.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284597 | BQ032998.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128044 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139222 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02792163 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02792175 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02792160 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02792171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02792160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03019090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02792175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02792171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22342395 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500092 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428364_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500091 | - | - |
| Mouse Genome 430 2.0 Array | 1428364_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428364_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69269_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69269_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019042.mm.1 | NM_001163573.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019042.mm.1 | NM_001163573.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019042.mm.1 | NM_027013.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019042.mm.1 | NM_027013.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019042.mm.1 | TC0300002465.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428364_PM_at | - | - |
| Mouse Expression Set 430 | 1428364_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428364_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428364_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428364_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428364_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 806567 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100360162 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100360162 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227006 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255227 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002107.1_66-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100360162 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100360162 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2787580 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227006 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2787580 | - | - |

Scnm1
Genomic Map