Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400025680.mm.1 |
TC0400004216.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025680.mm.1 |
NM_001285956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025680.mm.1 |
NM_001285958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025680.mm.1 |
NM_172874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025683.mm.1 |
TC0400004216.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025683.mm.1 |
NM_001285956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025683.mm.1 |
NM_001285958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025683.mm.1 |
NM_172874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035204.mm.1 |
TC0400004216.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400035204.mm.1 |
NM_001285956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035204.mm.1 |
NM_001285958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400035204.mm.1 |
NM_172874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025681.mm.1 |
TC0400004216.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025681.mm.1 |
NM_001285956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025681.mm.1 |
NM_001285958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025681.mm.1 |
NM_172874.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025684.mm.1 |
TC0400004216.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025684.mm.1 |
NM_001285956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025684.mm.1 |
NM_001285958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025684.mm.1 |
NM_172874.3 |
Refseq |