Assay Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153565.2 | NP_705793.1 | 10 - 11 | 2109 | 63 | |
| GenBank mRNA | AK043077.1 | - | 10 - 11 | 1958 | 63 | |
| AK149520.1 | - | 10 - 11 | 1888 | 63 | ||
| AY273821.1 | - | 10 - 11 | 1733 | 63 | ||
| BC038085.1 | - | 10 - 11 | 2124 | 4502199 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcsk9
Genomic Map