Assay Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153565.2 | NP_705793.1 | 9 - 10 | 1926 | 97 | |
| GenBank mRNA | AK043077.1 | - | 9 - 10 | 1775 | 97 | |
| AK149520.1 | - | 9 - 10 | 1705 | 97 | ||
| AY273821.1 | - | 9 - 10 | 1550 | 97 | ||
| BC038085.1 | - | 9 - 10 | 1941 | 4502199 | 97 | |
More Information
Set Membership: |
|

Pcsk9
Genomic Map