Assay Details
| Gene Symbol: Trp73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126330.1 | NP_001119802.1 | 4 - 5 | 828 | 81 | |
| NM_001126331.1 | NP_001119803.1 | 4 - 5 | 828 | 81 | ||
| NM_011642.3 | NP_035772.2 | 6 - 7 | 1118 | 81 | ||
| GenBank mRNA | AB221594.1 | - | 5 - 6 | 710 | 81 | |
| AK017412.1 | - | 3 - 4 | 359 | 81 | ||
| AK139633.1 | - | 4 - 5 | 829 | 81 | ||
| BC066045.1 | - | 4 - 5 | 715 | 6812399 | 81 | |
| Y19234.1 | - | 6 - 7 | 1119 | 81 | ||
| Y19235.1 | - | 4 - 5 | 822 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp73
Genomic Map