Assay Details
| Gene Symbol: Mfsd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178620.3 | NP_848735.1 | 9 - 10 | 814 | 82 | |
| GenBank mRNA | AK031384.1 | - | 9 - 10 | 1009 | 82 | |
| AK032043.1 | - | 9 - 10 | 1010 | 82 | ||
| BC016209.1 | - | 9 - 10 | 773 | 82 | ||
| BC079583.1 | - | 9 - 10 | 841 | 82 | ||
| BC088988.1 | - | 9 - 10 | 814 | 6418757 | 82 | |
| BC100547.1 | - | 9 - 10 | 2890 | 82 | ||
| BC137970.1 | - | 9 - 10 | 801 | 8860989 | 82 | |
| BC137971.1 | - | 9 - 10 | 802 | 8860990 | 82 | |
| BC145068.1 | - | 9 - 10 | 781 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd11
Genomic Map