Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100018509.mm.1 |
NM_178620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018509.mm.1 |
NM_178620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018509.mm.1 |
TC1100004282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018511.mm.1 |
NM_178620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018511.mm.1 |
NM_178620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018511.mm.1 |
TC1100004282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018514.mm.1 |
NM_178620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018514.mm.1 |
NM_178620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018514.mm.1 |
TC1100004282.mm.1 |
MTA TC |