Assay Details
| Gene Symbol: Mfsd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178620.3 | NP_848735.1 | 6 - 7 | 555 | 111 | |
| GenBank mRNA | AF015191.1 | - | 6 - 7 | 521 | 111 | |
| AK031384.1 | - | 6 - 7 | 750 | 111 | ||
| AK032043.1 | - | 6 - 7 | 751 | 111 | ||
| BC016209.1 | - | 6 - 7 | 514 | 111 | ||
| BC079583.1 | - | 6 - 7 | 554 | 111 | ||
| BC088988.1 | - | 6 - 7 | 555 | 6418757 | 111 | |
| BC100547.1 | - | 7 - 8 | 687 | 111 | ||
| BC137970.1 | - | 6 - 7 | 542 | 8860989 | 111 | |
| BC137971.1 | - | 6 - 7 | 543 | 8860990 | 111 | |
| BC145068.1 | - | 6 - 7 | 494 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd11
Genomic Map