Assay Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020569.3 | NP_065594.2 | 4 - 5 | 404 | 87 | |
| GenBank mRNA | AB015652.1 | - | 4 - 5 | 351 | 87 | |
| AK146368.1 | - | 4 - 5 | 316 | 87 | ||
| AK153948.1 | - | 4 - 5 | 361 | 87 | ||
| AK168341.1 | - | 4 - 5 | 334 | 87 | ||
| BC002187.1 | - | 4 - 5 | 310 | 3487763 | 87 | |
| CA456587.1 | - | 4 - 5 | 442 | 87 | ||
| CD776363.1 | - | 3 - 4 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Park7
Genomic Map