Assay Details
| Gene Symbol: Wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021499.2 | NP_067474.2 | 2 - 3 | 294 | 66 | |
| GenBank mRNA | AB034911.2 | - | 2 - 3 | 274 | 66 | |
| AK011773.1 | - | 2 - 3 | 258 | 66 | ||
| AK051688.1 | - | 2 - 3 | 238 | 66 | ||
| AK156969.1 | - | 2 - 3 | 293 | 66 | ||
| AK160331.1 | - | 2 - 3 | 266 | 66 | ||
| AK172690.1 | - | 2 - 3 | 293 | 66 | ||
| AK182927.1 | - | 1 - 2 | 169 | 66 | ||
| BC016120.1 | - | 2 - 3 | 257 | 66 | ||
| BY063723.1 | - | 2 - 3 | 295 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wrap73
Genomic Map