Assay Details
| Gene Symbol: Wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021499.2 | NP_067474.2 | 7 - 8 | 809 | 78 | |
| GenBank mRNA | AB034911.2 | - | 7 - 8 | 789 | 78 | |
| AK011773.1 | - | 7 - 8 | 772 | 78 | ||
| AK051688.1 | - | 7 - 8 | 753 | 78 | ||
| AK156969.1 | - | 7 - 8 | 808 | 78 | ||
| AK160331.1 | - | 7 - 8 | 781 | 78 | ||
| AK172690.1 | - | 7 - 8 | 808 | 78 | ||
| BC016120.1 | - | 7 - 8 | 772 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wrap73
Genomic Map