Assay Details
| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172095.1 | NP_001165566.1 | 17 - 18 | 2581 | 104 | |
| NM_144787.2 | NP_659036.1 | 17 - 18 | 2645 | 104 | ||
| GenBank mRNA | AK144103.1 | - | 17 - 18 | 2608 | 104 | |
| AK173023.1 | - | 17 - 18 | 2645 | 104 | ||
| BC020180.1 | - | 17 - 18 | 2587 | 4923943 | 104 | |
| BC042424.1 | - | 17 - 18 | 2581 | 5357211 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm4c
Genomic Map