Assay Details
| Gene Symbol: Ajap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099299.1 | NP_001092769.1 | 3 - 4 | 1601 | 71 | |
| GenBank mRNA | BC147229.1 | - | 3 - 4 | 965 | 9056280 | 71 |
| BC147230.1 | - | 3 - 4 | 965 | 9056273 | 71 | |
| BC147654.1 | - | 3 - 4 | 965 | 9087443 | 71 | |
| BC147658.1 | - | 3 - 4 | 965 | 9087447 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ajap1
Genomic Map