Assay Details
| Gene Symbol: Tnfrsf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033042.4 | NP_149031.2 | 3 - 4 | 682 | 70 | |
| GenBank mRNA | BC017526.1 | - | 3 - 4 | 682 | 4912933 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf25
Genomic Map