Assay Details
| Gene Symbol: Tnfrsf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291010.1 | NP_001277939.1 | 7 - 8 | 834 | 87 | |
| NM_033042.4 | NP_149031.2 | 4 - 5 | 784 | 87 | ||
| GenBank mRNA | AF329969.1 | - | 7 - 8 | 838 | 87 | |
| AK040632.1 | - | 4 - 5 | 1453 | 87 | ||
| BC017526.1 | - | 4 - 5 | 784 | 4912933 | 87 | |
More Information
Set Membership: |
|

Tnfrsf25
Genomic Map