Assay Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037717.3 | NP_001032806.2 | 9 - 10 | 768 | 144 | |
| GenBank mRNA | BC043932.1 | - | 2 - 3 | 95 | 144 | |
| BC157917.1 | - | 9 - 10 | 717 | 9007345 | 144 | |
| BC157928.1 | - | 9 - 10 | 717 | 9088101 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a6
Genomic Map