Assay Details
| Gene Symbol: Kctd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142731.1 | NP_001136203.1 | 2 - 3 | 1995 | 88 | |
| NM_134112.5 | NP_598873.2 | 2 - 3 | 311 | 88 | ||
| GenBank mRNA | AK054557.1 | - | 2 - 3 | 302 | 88 | |
| AY899808.1 | - | 2 - 3 | 311 | 88 | ||
| BC087623.1 | - | 2 - 3 | 210 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kctd1
Genomic Map