Assay Details
| Gene Symbol: Kctd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142731.1 | NP_001136203.1 | 3 - 4 | 2134 | 143 | |
| NM_134112.5 | NP_598873.2 | 3 - 4 | 450 | 143 | ||
| GenBank mRNA | AK016623.1 | - | 2 - 3 | 495 | 143 | |
| AY899808.1 | - | 3 - 4 | 450 | 143 | ||
| BC022615.1 | - | 1 - 2 | 118 | 143 | ||
| BC087623.1 | - | 3 - 4 | 349 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kctd1
Genomic Map