Assay Details
| Gene Symbol: Trp73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126330.1 | NP_001119802.1 | 1 - 2 | 280 | 58 | |
| NM_001126331.1 | NP_001119803.1 | 1 - 2 | 280 | 58 | ||
| GenBank mRNA | AK139633.1 | - | 1 - 2 | 281 | 58 | |
| BC066045.1 | - | 1 - 2 | 167 | 6812399 | 58 | |
| Y19235.1 | - | 1 - 2 | 274 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp73
Genomic Map